Foam Pistol Bottle, Nito Clean
Lieferant: WERNER MERTZ
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0705727
141-0285EA 169 EUR InStock
141-0285
Foam Pistol Bottle, Nito Clean
Flaschen Sprühflaschen
Foam gun for applying a stable foam carpet on horizontal and vertical surfaces. The dosage can be adjusted.
For use with cleaning agents. Can be connected to a standard water tap.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge