Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cell disruptors, Sonifier® SFX series

Lieferant: BRANSON

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

101-063-963R 101-063-966R 101-063-970R 101-063-971R
BRAN101-063-963REA 4456 EUR InStock
BRAN101-063-963R 142-0143 142-0145 142-0144
Cell disruptors, Sonifier® SFX series
Homogenisatoren Ultraschall-Desintegratoren
A series of sonifiers with advanced sample processing capabilities and a digital, intuitive menu driven interface.

  • Continuous or pulsed ultrasonics
  • Ergonomic handheld converter with recessed push button and LED indicator
  • Energy mode for delivery of a precise user-determined input of energy (J)
  • On screen process monitoring of power level, energy usage, sample temperature and time
  • Sample processing programs - up to 20 user defined programs
  • Amplitude control range from 10 to 100% ( use of microtips limit this to 70%)

SFX150: Designed for low volume samples, the SFX150 is quiet enough to operate in on a lab bench without a sound enclosure making it ideal for use with a handheld converter.

SFX250 and SFX 550: These sonifiers feature improved ease of use, precision, and repeatability with advanced energy and temperature control modes, process monitoring capabilities, and programming features.

Advanced energy control mode delivers precise energy input in continuous or pulse modes
True temperature control manages sample temperature within user-specified limits (requires separate temperature probe)
Control modes include time, temperature, and energy
High efficiency, stand-mountable converter requires no internal cooling
Scrollable, multi-function screen with pushbuttons
In-process feedback displays experimental progress together with key variables such as power level, energy usage, and sample temperature.

Advanced energy control mode delivers precise energy input in continuous or pulse modes.
True temperature control manages sample temperature within user-specified limits (requires separate temperature probe).
Control modes include time, temperature, and energy
High efficiency, stand-mountable converter requires no internal cooling
Scrollable, multi-function screen with pushbuttons.
In-process feedback displays experimental progress together with key variables such as power level, energy usage, and sample temperature.
Amplitude control range from 10 - 100% ( use of microtips limit this to 70%)

Achtung! Bitte beachten: Never use a probe with replacement tip with low surface tension liquids.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge