Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Aluminum silicate

Lieferant: ALFA AESAR

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


14231.36 14231.A3
14231.36EA 48.8 EUR InStock
14231.36 14231.A3
Aluminum silicate
Aluminum silicate
Aluminum silicate is used to prepare aluminum-silicate polymer composite (PASiC), which is an inorganic coagulant used in water treatment. It is used as a catalyst for the pyrolysis of rice husk to crude bio -oil, which upgraded to supercritical ethanol. Further, it is used as filler in paper, rubber and in paints.

CAS No.: 12428-46-5 Molecular Formula: Al2Si2O7.2H2O Formula Weight: 258.16 (222.13anhy) Physical Form: Powder Appearance: White to pale grey Odour: Odourless MDL No.: MFCD00062311


Formel: Al₂Si₂O₇·2H₂O
MW: 258.16 g/mol (222.13 anhy)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00062311
CAS-Nummer: 12428-46-5
Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge