14534.36
14534.A3
14534.36EA
96.1
EUR
InStock
14534.36
14534.A3
Ammoniumsulfamat ≥98% ACS
Ammoniumsulfamat
Purity: 98% min
CAS No.: 7773-06-0
Molecular Formula: H6N2O3S
Formula Weight: 114.12
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Odour: Weak
pH: 5.2
Melting Point: 131-135°
Boiling Point: 160° dec.
Solubility: Very soluble in water, liquid ammonia. Slightly soluble in ethanol, glycerol, glycol
Merck Reference: 14,554
MDL No.: MFCD00011429
In electroplating solutions, making nitrous oxide gas, for manufacture of flameproofing textiles and paper products