ampuled under argon
14552.06
14552.14
14552.22
14552.06EA
87.2
EUR
InStock
14552.06
14552.14
14552.22
Aluminiumchlorid, dry_ultra 99,99 Metals basis
Aluminiumchlorid
Purity: 99.99%
CAS No.: 7446-70-0
Molecular Formula: AlCl3
Formula Weight: 133.34
Physical Form: Powder, Ampouled under argon
Density: 2.44
pH: 2,4
Melting Point: 190° subl.
Merck Reference: 14,337
UN No.: 1726
MDL No.: MFCD00003422