14596.09
14596.09EA
178
EUR
InStock
14596.09
Terbium(III)sulfat Octahydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Terbium(III)sulfat Octahydrat
Purity: 99.9%
CAS No.: 13842-67-6
Molecular Formula: Tb2(SO4)3.8H2O
Formula Weight: 750.19 (606.05anhy)
Physical Form: Crystalline
Melting Point: 360° -8H2O
MDL No.: MFCD00149872