14891.09
14891.18
14891.30
14891.09EA
42.4
EUR
InStock
14891.09
14891.18
14891.30
Ethansulfonsäure ≥96%
Ethanesulphonic acid
Purity: 96%
CAS No.: 594-45-6
Molecular Formula: CH3CH2SO3H
Formula Weight: 110.13
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 1.343
Refractive Index: 1.4350
Appearance: Colourless to yellow
Melting Point: -17°
Boiling Point: 122-123°/0.01mm
Beilstein Reference: 1743071
UN No.: 2586
MDL No.: MFCD00007529