Brushes, swivel-head, for flasks
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
K-2500
149-1210EA 28.7 EUR InStock
149-1210
Brushes, swivel-head, for flasks
Bürsten
For intensive and gentle cleaning. Fits flasks from 500 to 5000 ml with SGJ/NS 29 openings.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge