Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Isopentylnitrit (Isomerengemisch) 96%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ200

150495-100ML 150495-500ML 150495-5ML
150495-100MLEA 69.5 EUR InStock
150495-100ML 150495-500ML 150495-5ML
Isopentylnitrit (Isomerengemisch) 96%, Sigma-Aldrich®
Isopentylnitrit (Isomerengemisch)
Isopentyl nitrite, also known as isoamyl nitrite, is an oxidant, and nitrosating agent for phosphoramidites which rearrange into phosphotriesters during nitrosation.

  • Isopentyl nitrite has been used as a radical initiator to functionalize multi-walled carbon nanotubes
  • As a reagent in the preparation of 4-carboxy-phenyl diazonium salt tetrafluoroborate
  • In the synthesis of peroxynitrite

Isopentyl nitrite, also known as isoamyl nitrite, is used for the generation of benzyne from anthranilic acid. It is a smooth muscle relaxant that may be suitable as nitrovasodilators and as recreational drugs known as poppers. Its applications are also seen in kits to treat cyanide poisoning, its function being to oxidise hemoglobin (Hb) to methemoglobin (metHb) which reacts with cyanide to form the non-toxic cyano-metHb. It is used a diagnostic assay manufacturing, hematology, histology.


Formel: C₅H₁₁NO₂
MW: 117.15 g/mol
Siedepunkt: 99 °C
Melting Pt: ≤80 °C
Dichte: 0,872 g/ml (25 °C)
MDL Number: MFCD00002057
CAS-Nummer: 110-46-3
EINECS: 203-770-8
UN: 1113
ADR: 3 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07b0a40203d4f9eb2536f5e13063c9cd4bfae4bc&vid=07b0a40203d4f9eb2536f5e13063c9cd4bfae4bc&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Colorless to Yellow and Colourless to Green-Yellow
Appearance (Form) Liquid
Purity (GC AREA %) ≥95.5 %
Infrared spectrum Conforms to structure

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge