Organic reference standard, Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, mixture of 3 isomers ortho-ortho, ortho-para, para-para, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
15139-250MG
15139-250MGEA 210 EUR InStock
15139-250MG
Organic reference standard, Bisphenol F bis(3-chloro-2-hydroxypropyl) ether, mixture of 3 isomers ortho-ortho, ortho-para, para-para, Supelco®
Standards Organische Standards
Bisphenol F bis(3-chloro-2-hydroxypropyl) ether a additive used during synthesis of organosol resin preventing theremal degradation of polymer. During the removal of hydrochloric acid in organosol resin results in this chlorinated compound.
  • Used as reference standard in RP-HPLC-APCI-MS-MS analysis for optimising and quantifying pressurised liquid extraction for BFDGE and its derivatives from canned food
Standard for determining toxic monomers released from polymers of the inner coating of cans.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52538f24a14c469801d093ed2fbff37f841b959f&vid=52538f24a14c469801d093ed2fbff37f841b959f&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge