Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Acridinorange (Zinkchlorid-Doppelsalz) 85% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ200

158550-100G 158550-10G 158550-25G
158550-100GEA 460 EUR InStock
158550-100G 158550-10G 158550-25G
Acridinorange (Zinkchlorid-Doppelsalz) 85% (Farbstoffgehalt), Sigma-Aldrich®
Acridinorange (Zinkchlorid-Doppelsalz)
Acridine orange hemi (zinc chloride) salt has been used to stain primary human umbilical vein endothelial cells (HUVEC) and is used in micronucleus tests (MNTs). Acridine orange, a cell-permeable metachromatic fluorescent cationic dye that intercalates DNA and RNA, is used in fluorescence and epiflouresence microscopy. Acridine orange dye has been used to analyse mitochondria and lysosomal content by flow cytometry, characterise multidrug resistance, and measure changes in mitochondrial mass during apoptosis in rat thymocytes.

Acridine orange (AO) is a fluorescent dye with two different staining characteristics. In fixed tissues, it acts as a metachromatic dye and differentially stains DNA (green) and RNA (orange). But, in living cells it acts as a pH indicator and produce brilliant orange-red in acidic condition. AO has been used to identify apoptotic cells in drosophila embryos. Acridine orange staining method has been used to deliver more consistent data in the micronucleus test. It is used a diagnostic assay manufacturing, hematology, histology.


Formel: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
Storage Temperature: Room Temperature
MDL Number: MFCD00081043
CAS-Nummer: 10127-02-3
EINECS: 233-353-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92df35e7d21393cca5f51cfdc019e7c10016059e&vid=92df35e7d21393cca5f51cfdc019e7c10016059e&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength 265 - 271 nm
Extinction Coefficient ≥38000
Carbon ≥46.3%
Nitrogen ≥9.5%
ICP Major Analysis Zinc Component Confirmed
Solubility (Turbidity) Clear
Solubility (Colour) Orange to very dark orange and brown to very dark brown and red-orange and red-brown and orange-brown and very dark red-brown

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge