Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Gel filtration media, Sephadex™ G-50

Lieferant: GE HEALTHCARE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


17-0041-01 17-0042-01 17-0042-02 17-0043-01 17-0043-02
17-0041-01EA 557 EUR InStock
17-0041-01 17-0042-01 17-0042-02 17-0043-01 17-0043-02
Gel filtration media, Sephadex™ G-50
Protein-Aufbereitungs Protein Purification Resins
Sephadex™ G-50 Medium is well established gel filtration medium for desalting and buffer exchange of biomolecules >30000 molecular weight. The larger bead size of G-50 Medium is suited for large scale separations.

  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step
  • Suitable for purification of DNA from small molecules by gel filtration
  • Larger bead size for large scale group separations where high flow rates and low operating pressures are required
  • Classic gel filtration medium

Sephadex™ is a gel filtration resin prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex™ differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex™ G-50 is one of six different G-types ranging from G-10 for small molecules to G-100 for very large molecules. Sephadex™ G-50 is available in 3 different particle sizes (Medium, Fine and Superfine). Superfine has the smallest bead size for higher efficiency with shorter diffusion distances. Medium is preferred for large scale group separations where high flow rates and low operating pressures are required.

Particle size (wet): 17 to 70 µm (Superfine) / 40 to 160 µm (Fine) / 100 to 300 µm (Medium).


Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge