Gelfiltrationsmedien, Sephadex™ LH-20
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0090-01 17-0090-02 17-0090-10
17-0090-01EA 587 EUR InStock
17-0090-01 17-0090-02 17-0090-10
Gelfiltrationsmedien, Sephadex™ LH-20
Protein-Aufbereitungs Protein Purification Resins
Sephadex™ LH-20 is a liquid chromatography media for molecular sizing of natural product such as steroid, terpenoid, lipid and low molecular weight peptides. Sephadex™ LH-20 is pre-pared by hydroxypropylation of Sephadex™ G-25, a bead-formed dextran medium.
  • Designed to use in polar organic solvents and aqueous solvent mixtures
  • Excellent for fractionation of lipids, hormones, vitamins, and other small biomolecules
  • Made from hydroxypropylated dextran beads that have been cross-linked to yield a polysaccharide network
  • Powerful chromatographic selectivity due to dual hydrophilic and lipophilic nature of the matrix
  • Excellent batch-to-batch reproducibility
Sephadex™ LH-20 is useful at both analytical and industrial scale for the preparation of closely related molecular species. Due to the physico-chemical properties of this media, it can be used either during initial purification prior to polishing by high performance ion exchange or reversed phase chromatography or as the final polishing step e.g. during the preparation of diastereomers.
Sephadex LH-20 is prepared by hydroxypropylation of Sephadex G-25, a bead-formed dextran medium.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge