Sepharose™ CL-4B Gel Filtration Base Matrix
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0150-01
17-0150-01EA 1090 EUR InStock
17-0150-01
Sepharose™ CL-4B Gel Filtration Base Matrix
Protein-Aufbereitungs Protein Purification Resins
Sepharose™ CL-4B is a cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix.
  • 4% cross-linked agarose gel filtration media
  • Proven base matrix for coupling affinity ligands
  • Higher degree of crosslinking improve matrix stability
The matrix is not pre-activated and the user performs all steps in coupling.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge