Affinity chromatography media, Lentil Lectin Sepharose™ 4B
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0444-01
17-0444-01EA 862 EUR InStock
17-0444-01
Affinity chromatography media, Lentil Lectin Sepharose™ 4B
Protein-Aufbereitungs Protein Purification Resins
Lentil lectin is a metalloprotein, containing Ca₂₊ and Mn₂₊, which binds reversibly to polysaccharides and glycoconjugates containing glucose or mannose type sugars.
  • For affinity chromatography
  • Cyanogen bromide activation coupling methods
  • Stable to all commonly used aqueous buffers
  • Chelating agents should be avoided
  • Negligible volume variation due to changes in pH or ionic strength
Lentil Lectin Sepharose 4B is a generally applicable group specific adsorbent and is routinely used in the preparation and purification of a number of glycoproteins and carbohydrate containing molecules. Binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues. Compared to Con A, also for glycoprotein purification, Lentil Lectin distinguishes less sharply between glucosyl and mannosyl. Retains binding in 1% deoxycholate and is especially valuable for purification of detergent-solubilized membrane proteins. Chelating agents such as EDTA, 8 M urea or solutions having a pH below 3 should be avoided as these conditions result in removal of manganese from the lectin and loss of activity.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..