pI markers
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0471-01 17-0473-01 29-4435-48
17-0471-01EA 439 EUR InStock
17-0471-01 17-0473-01 29-4435-48
pI markers
Elektrophorese-Reagenzien Molekulargewichtsmarker
pI markers are lyophilized mixtures of stable, salt-free, highly purified proteins.
  • To be used for isoelectric focusing (IEF)
The precisely defined pI of the markers allows a permanent and accurate calibration of sample components. pH gradients and pI can be measured to within 0.01 to 0.05 pH unit. The progress of the focusing experiment can be monitored visually by the focusing of methyl red or cytochrome c.
The markers may be run in parallel with the experimental samples on isoelectric focusing gels. These markers are not prestained.
Bestellinformation: The broad-range pI markers kit contains 12 amylglucosidase markers, carbonic anhydrase B from both bovine and human sources, beta-lactoglobulin A, lentil lectin for acidic, middle and basic bands, methyl red, myoglobin for both acidic and basic bands, trypsin inhibitor and trypsinogen. The high-range kit contains 10 amylglucosidase markers, carbonic anhydrase B from both bovine and human sources, Cytochrom C, beta-lactoglobulin A, lentil lectin for acidic, middle and basic bands, myoglobin for both acidic and basic bands and trypsinogen. Kits are not for use in denaturing (urea) systems.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge