Superose™ 12 Gel Filtration Media, Prep Grade
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0536-01
17-0536-01EA 760 EUR InStock
17-0536-01
Superose™ 12 Gel Filtration Media, Prep Grade
Protein-Aufbereitungs Protein Purification Resins
Good resolution separations of proteins and other biomolecules according to size, Wide separation range – separations in two molecular weight ranges from 1,000 up to 5 million, Excellent reproducibility and durability
  • Excellent reproducibility and durability
Achieve high-resolution separations across exceptionally broad molecular-weight ranges. Low ionic strength eluents (< 0.05 M) can utilize a hydrophobic interaction component to alter the selectivity of Superose for some lipids, peptides, and small aromatic compounds. High performance gel filtration media specially designed for preparative purification of biomolecules. Superose 6 is designed for high-resolution chromatography. Superose 6 prep grade permits easy, efficient scale-up to preparative separations. Superose 12 is for high resolution purification of proteins from 1 × 103 to 3 × 105 molecular weight, while Superose 12 prep grade is useful for preparative purification of these higher molecular weight proteins.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge