Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

DNA clean up, Sephadex® G-50 DNA Grade F

Lieferant: GE HEALTHCARE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


17-0573-02 17-0573-01
17-0573-02EA 418 EUR InStock
17-0573-02 17-0573-01
DNA clean up, Sephadex® G-50 DNA Grade F
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Sephadex G-50 grade F is a size exclusion chromatography resin designed for desalting and buffer exchange, and for purification of DNA and oligonucleotides greater than 20 bases in length using spin columns.

  • For purification of DNA from small molecules by size exclusion
  • Tested to ensure reproducibly high recovery of DNA
  • Sephadex G-50 DNA Grade is used in various G-50 columns, such as AutoSeq G-50 columns, ProbeQuant G-50 Micro Columns, MicroSpin G-50 Columns, and NICK Columns
  • Suitable for researchers who prefer to prepare their own columns for purifying nucleic acids
  • Excellent for rapid purification of newly synthesized oligonucleotides > 10-mers, including oligonucleotide probes, in 100 to 150 µl of deprotection solution using spin-column chromatography
  • Useful for desalting or buffer exchange of DNA and removal of unincorporated radionucleotides from end-labeled oligonucleotides (at least 10 bases in length) in a volume of 10 to 100 µl

Sephadex G-50 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex G-50 is a well-established gel filtration resin for desalting and buffer exchange of biomolecules > 30 000 molecular weight, and with a spin protocol can be used for DNA and oligo purification of molecules greater than 20 bases in length.

Parameters
Particle Size Dry: 50 µm to 150 µm
Application: For purification of DNA from small molecules by gel filtration
Fractionation range (Mr) (globular proteins): 1500 to 30000
Fractionation (Mp) Dextrans: 500 to 10000
Fractionation Range (Mr) DNA Exclusion Limit: 20


Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge