Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

DNA clean up, Sephadex® G-100 DNA grade

Lieferant: GE HEALTHCARE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


17-0574-02
17-0574-02EA 531 EUR InStock
17-0574-02
DNA clean up, Sephadex® G-100 DNA grade
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Sephadex™ G-100 DNA Grade is ideal for use in preparing spin columns for DNA purification.

  • For DNA purification from small molecules by gel filtration in spin columns with an exclusion limit of 25bp ds DNA
  • Tested to ensure reproducibly high recovery of DNA
  • Suitable for researchers who prefer to prepare their own columns for nucleic acid purification

Sephadex G-100 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Spin columns packed with Sephadex G-100 DNA Grade allow for high-recovery nucleic acid purification via gel filtration. Gel filtration resins do not exhibit a fixed exclusion limit when used in a spin-column format. In spin-column chromatography, the observed exclusion properties that allow the product to pass through the gel while the smaller impurities are retained depend on experimental factors, such as the resin used, sample volume, product size, and the g-forces used in the purification process.


Parameter
Particle Size Dry: 20 µm-50 µm
Pressure/Flow Specification: 0.096 bar
Fractionation range (Mr) (globular proteins): 4000–100000
Fractionation (Mp) Dextrans: 1000-100000


Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge