Affinity chromatography media, Chelating Sepharose™ Fast Flow
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0575-01 17-0575-02
17-0575-01EA 520 EUR InStock
17-0575-01 17-0575-02
Affinity chromatography media, Chelating Sepharose™ Fast Flow
Protein-Aufbereitungs Protein Purification Resins
Chelating Sepharose Fast Flow is composed of cross-linked 6% agarose beads modified with iminodiacetic immobilized to the base matrix by stable ether linkages and sufficiently long spacer arms.
  • Uncharged IMAC resin allowing possibility to charge with your metal ion of choice for optimized selectivity
  • Convenient purification of native and recombinant proteins
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Stable in commonly used aqueous buffers - 1 M NaOH, 20% ethanol, 8 M guanidine hydrochloride, 8 M urea, 0.01 M HCl, non-ionic detergents, acetate buffer pH 4
Chelating Sepharose Fast Flow is available in a range of different bulk pack sizes for easy scale-up and process development. As member of the BioProcess media range, Chelating Sepharose Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
As BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support. Chelating Sepharose Fast Flow is supplied free of metal ions, allowing the user to charge it with the most appropriate metal ion for purification of a target protein.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge