Ion exchange chromatography media, DEAE Sepharose™ Fast Flow
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0709-01 17-0709-10
17-0709-01EA 754 EUR InStock
17-0709-01 17-0709-10
Ion exchange chromatography media, DEAE Sepharose™ Fast Flow
Protein-Aufbereitungs Protein Purification Resins
DEAE Sepharose™ Fast Flow is part of the Sepharose™ Fast Flow ion exchange platform. It is composed of cross-linked 6% agarose beads, with diethylaminoethyl (DEAE) weak anion exchange groups. Scale-up with DEAE Sepharose™ Fast Flow is straight forward.
  • Well-proven weak anion exchanger developed for industrial downstream processes
  • Used extensively for capture and intermediate purification of a wide range of approved biopharmaceuticals
  • High chemical stability allows for well proven CIP and sanitisation protocols
  • The hydrophilic nature of the base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
DEAE Sepharose Fast Flow has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols. Scale-up with DEAE Sepharose Fast Flow is straight forward and the medium is available in a range of pre-packed process development tools. As member of the BioProcess media range, DEAE Sepharose Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
DEAE Sepharose Fast Flow is part of the Sepharose Fast Flow ion exchange platform, which has been the industrial standard for ion exchange chromatography during recent decades. It is composed of crosslinked 6% agarose beads, with diethylaminoethyl (DEAE) weak anion exchange groups.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge