Affinity chromatography columns, HiTrap™ NHS-Activated HP
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0716-01 17-0717-01
17-0716-01EA 266 EUR InStock
17-0716-01 17-0717-01
Affinity chromatography columns, HiTrap™ NHS-Activated HP
Chromatographiesäulen
HiTrap NHS-activated HP prepacked chromatography columns allow you to design an affinity column suitable for your purification needs.
  • Easy coupling of any ligand containing primary amine groups
  • Packed with NHS-Activated Sepharose High Performance resin
  • Convenient prepacked columns can be used with a syringe, pump, or chromatographic system, such as ÄKTA systems
This NHS-activated resin can be used with any primary amine-containing ligand. The resin reacts with primary amine groups to form stable amide linkages that covalently immobilize antibodies or other proteins for affinity purification. The resin enables simple coupling, carried out at close to neutral pH.
The NHS-activated Sepharose High Performance (HP) resin in these columns is delivered in 100% isopropanol to protect the active groups. It contains ~ 10 μmol NHS groups/mL resin.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge