Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Size exclusion chromatography columns, illustra™ NAP™

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

17-0852-01 17-0852-02 17-0853-01 17-0853-02 17-0854-01 17-0854-02
17-0852-01EA 294 EUR InStock
17-0852-01 17-0852-02 17-0853-01 17-0853-02 17-0854-01 17-0854-02
Size exclusion chromatography columns, illustra™ NAP™
Chromatographiesäulen
NAP™ columns are disposable columns prepacked with Sephadex™ G-25 DNA grade and require only gravity to run.

  • For rapid and efficient purification of DNA and oligonucleotides (>10-mers) in less than 15 minutes by gravity flow
  • Prepacked with Sephadex™ G-25 DNA grade in distilled water containing 0,15% Kathon CG/ICP Biocide
  • Available in three sizes depending on sample volume: 0,5 ml (NAP™-5), 1 ml (NAP™-10) or 2,5 ml (NAP™-25)
  • Useful for small-scale purification, desalting and buffer exchange

Lieferumfang: Each package includes a complete step-by-step protocol describing the fixed volumes needed for equilibration, washing and sample elution.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge