Protein A
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0872-50
17-0872-50EA 951 EUR InStock
17-0872-50
Protein A
Proteine und Peptide
Staphylococcal protein A is the classical immunoglobulin-binding reagent for a wide range of antibodies, used extensively in affinity chromatography and antibody detection applications. The protein is enterotoxin-free and gives reliable results. Protein A binds the Fc regions of IgG antibodies from a variety of species, for convenient antibody purification.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge