Affinity chromatography media, IMAC Sepharose™ High Performance
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0920-07
17-0920-07EA 1471 EUR InStock
17-0920-07
Affinity chromatography media, IMAC Sepharose™ High Performance
Protein-Aufbereitungs Protein Purification Resins
IMAC Sepharose High Performance media is easily charged by the user with the metalion of choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent purification of polyhistidine tagged proteins.
  • For optimizing purification of histidine-tagged proteins when Ni2+ is not the best choice of metal ion
  • Conveniently charge with your metal of choice
  • 34 μm bead size for high performance purification
  • High binding capacity
  • Stable in commonly used aqueous buffers
  • Avoid using chelating agents, e.g. EDTA, EGTA, citrate
Conveniently charge with your metal of choice. IMAC Sepharose FF is supplied free of metal ions. It is charged by the user with the transition metal ion of choice (e.g. Cu₂₊, Zn₂₊, Ni₂₊, or Co₂₊); these metal ions will bind to the covalently immobilized chelating ligand on the Sepharose. Small bead size for high performance purification and high binding capacity.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge