Hydrophobic interaction chromatography columns, HiTrap™ Butyl-S-FF Sepharose™
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0978-13
17-0978-13EA 245 EUR InStock
17-0978-13
Hydrophobic interaction chromatography columns, HiTrap™ Butyl-S-FF Sepharose™
Chromatographiesäulen
HiTrap™ Butyl-S-FF are prepacked Butyl-S Sepharose™ 6 Fast Flow columns for hydrophobic interaction chromatography (HIC).
  • Butyl-S Sepharose™ 6 Fast Flow is well-established and is the least hydrophobic medium in the Sepharose™ Fast Flow HIC range
  • Can be used with syringes, single laboratory pump or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap™ columns in series or using HiPrep™ columns
The columns are used for small-scale protein purification as well as for screening of binding and elution conditions.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge