Affinity coupling media, CNBr-activated Sepharose™ 4
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-0981-01
17-0981-01EA 469 EUR InStock
17-0981-01
Affinity coupling media, CNBr-activated Sepharose™ 4
Protein-Aufbereitungs Protein Purification Resins
CNBr-activated Sepharose™ 4 Fast Flow is based on the Sepharose™ Fast Flow platform. The resin is composed of cross-linked 4% agarose beads that has been pre-activated with cyanogen bromide.
  • CNBr-activated BioProcess medium designed for coupling of large amino-containing ligands
  • Easy, rapid and efficient coupling
  • This resin allows for multi-point attachment of protein ligands which minimizes ligand leakage
  • BioProcess medium supported for industrial applications and well-established in approved processes
The resin is composed of cross-linked 4% agarose beads that has been pre-activated with cyanogen bromide. CNBr-activated Speharose 4 Fast Flow is designed for multipoint attachment of protein ligands containing amino groups. The preparation and use of affinity chromatography media by coupling biospecific ligands to CNBr-activated Sepharose 4 Fast Flow is a widely used, successful and well-documented approach, with an easy, rapid and efficient coupling procedure. CNBr-activated Sepharose 4 Fast Flow is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As member of the BioProcess media range, CNBr-activated Sepharose 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
CNBr-activated Sepharose 4 Fast Flow is based on the well established Sepharose Fast Flow platform.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge