Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Desalting columns with Sephadex G-25 resin, HiTrap™

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

17-1408-01 29-0486-84
17-1408-01EA 310 EUR InStock
17-1408-01 29-0486-84
Desalting columns with Sephadex G-25 resin, HiTrap™
Chromatographiesäulen
HiTrap Desalting are ready-to-use 5 ml columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange.

  • Convenient desalting and buffer exchange of sample volumes up to 1,5 ml
  • Two to five columns can be connected in series for sample sizes from 3 ml to 7,5 ml
  • Replaces dialysis for desalting protein, which saves time and reduces labor
  • Requires less than one minute with typically > 95% recovery
  • HiTrap column format is convenient for use with a syringe, a peristaltic pump, or a chromatography system

HiTrap Desalting is a prepacked, ready-to-use column for group separation between high and low molecular weight substances. Applications include buffer exchange prior to or after different chromatographic steps, removal of low molecular weight contaminants, or removal of reagents to terminate a reaction.

The special design of the column, together with the well-known resin Sephadex G-25 Superfine, provides fast and reproducible separations in a convenient format.

The exclusion limit is approximately Mr 5000. This ensures group separation of proteins/peptides larger than Mr 5000 from smaller molecules.

Separations are easily performed with a syringe, a pump, or a chromatography system such as an ÄKTA system.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge