Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Affinity chromatography columns, HisTrap™ excel

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

17-3712-05 17-3712-06 29-0485-86
17-3712-05EA 270 EUR InStock
17-3712-05 17-3712-06 29-0485-86
Affinity chromatography columns, HisTrap™ excel
Chromatographiesäulen
HisTrap excel columns are prepacked with Ni Sepharose excel affinity resins for capture and purification of histidine-tagged proteins secreted into eukaryotic cell culture supernatants by immobilized metal ion affinity chromatography (IMAC).

  • Load eukaryotic cell culture samples containing secreted histidine-tagged proteins directly with retained binding capacity
  • Increase target protein yield and decrease degradation through reduced and simplified sample handling
  • HisTrap excel columns allow direct purification of cell-free, unclarified samples

HisTrap excel 1 ml and 5 ml are ready-to-use IMAC columns prepacked with Ni Sepharose excel. The design of the columns in combination with the specific properties of the resin enables fast and convenient purifications.

The special type of filter in the top and bottom of the columns makes it possible to load large volumes of cell-free, unclarified samples directly on the columns without causing back pressure problems. This time-saving property helps prevent degradation and loss of sensitive target proteins.

HisTrap excel columns are made of biocompatible polypropylene.

HisTrap excel columns can be operated either with a peristaltic pump or with chromatography systems such as ÄKTA systems. The columns cannot be opened or refilled.

Lieferumfang: Supplied with a stopper on the inlet and a snap-off end on the outlet.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge