Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cation exchange chromatography column, HiScreen™ Capto™ S ImpAct

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

17-3717-47
17-3717-47EA 163 EUR InStock
17-3717-47
Cation exchange chromatography column, HiScreen™ Capto™ S ImpAct
Chromatographiesäulen
HiScreen™ Capto™ S ImpAct columns are prepacked with Capto™ S ImpAct cation exchange chromatography medium. The columns are an excellent choice for method optimisation and parameter screening.

  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Easily connected in series to achieve 200 mm bed height
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
  • Strong cation exchange BioProcess medium, designed for immediate purification and polishing of a wide range of biomolecules, especially monoclonal antibodies
  • Flow rate of 1,2 to 2,3 ml/min

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge