Hydrophobic interaction chromatography column, HiScreen™ Capto™ Butyl ImpRes
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-3719-10
17-3719-10EA 219 EUR InStock
17-3719-10
Hydrophobic interaction chromatography column, HiScreen™ Capto™ Butyl ImpRes
Chromatographiesäulen
HiScreen™ Capto™ Butyl ImpRes is a ready-to-use column prepacked with Capto™ Butyl ImpRes hydrophobic interaction chromatography resin.
  • Excellent for method optimisation and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge