Q Sepharose™ XL Ion Exchange Chromatography Media, Cytiva
Lieferant: GE HEALTHCARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
17-5072-01
17-5072-01EA 764 EUR InStock
17-5072-01
Q Sepharose™ XL Ion Exchange Chromatography Media, Cytiva
Protein-Aufbereitungs Protein Purification Resins
Q Sepharose XL is based on a highly cross-linked, bead formed 6% agarose matrix similar to the well established Q Sepharose Fast Flow media.
  • High loading capacities give more productive capture from clarified feedstocks
  • Strong anion exchanger designed for capture steps
  • High chemical stability for effective CIP/sanitization
The hydrophilic nature of the base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool. With its high loading capacities and excellent physical and chemical stability, Q Sepharose XL is suitable for the capture stage of a downstream process, which is characterised by large loads of crude samples. As member of the BioProcess media range, Q Sepharose XL meets industrial demands with security of supply and comprehensive technical and regulatory support.
Dextran chains are covalently coupled to the agarose matrix and modified with quaternary ammonium (Q) strong anion exchange groups through chemically stable ether bonds. The stable agarose matrix and long, flexible chains of dextran with bound charged groups work together to increase loading capacity whilst allowing high flow rates.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..