Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cation exchange chromatography column, HiScreen™ Capto™ SP ImpRes

Lieferant: Cytiva

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

17-5468-15
17-5468-15EA 163 EUR InStock
17-5468-15
Cation exchange chromatography column, HiScreen™ Capto™ SP ImpRes
Chromatographiesäulen
HiScreen Capto SP ImpRes column is packed with strong cation exchange modern resin for fast high resolution protein purification in process development.

  • Prepacked with Capto SP strong cation exchange chromatography resin, suitable for intermediate protein purification and large-scale polishing
  • Convenient method optimization and parameter screening because of the 10 cm bed height
  • These columns are easily connected in series to increase bed height to 20 cm
  • Fast results and minimal consumption of sample and buffer because of the small bed volume
  • BioProcess columns packed with the same chromatography resin and using the same linear fluid velocity can produce scalable, reproducible results

High resolution ion exchange chromatography column when a strong cation exchanger is required. The Capto SP ImpRes base matrix creates good pressure and flow characteristics, which in turn deliver high resolution results in both purification and polishing.

Both the column format and SP resin are highly effective in parameter and method optimization and robustness testing, particularly during the development of a new purification process.

With a small 4,7 ml volume and 10 cm bed height, this HiScreen column can be used to perform scalable experiments at relevant process flow rates. When necessary, two columns can be connected in series to achieve a 20 cm bed height.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge