Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ionenaustauscherpatronen

Lieferant: AQUADEM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

REGE800S DEM-E100-001 DEM-E200-001 DEM-E300-001 DEM-E500-001 DEM-E600-001 E080 E200 E300 E500 E600 PMEFP210 PMEFP310 REGE100S REGE200S REGE300S REGE500S REGE600S E100
AQUDREGE800SEA 0 EUR InStock
AQUDREGE800S 171-0100 171-0101 171-0102 171-0103 171-0104 171-0106 171-0107 171-0108 171-0109 171-0110 171-0122 171-0123 AQUDREGE100S AQUDREGE200S AQUDREGE300S AQUDREGE500S AQUDREGE600S 171-0337
Ionenaustauscherpatronen
Wasseraufbereitungssysteme Ionenaustauscher
The polyamide range is a set of demineralisation columns using mixed-bed ion exchange resin technology. These devices ensure deionised water production of between 5 and 15 MΏ/cm according to whether the installation type is single or double cycle.

  • L×D×H: 173×102×663 mm
  • Connections: ¾ inch.

Connections: ¾ inch.

These filtration panels are accessories for demineralisation cylinders specifically designed for use in laboratories or health facilities. The cylinders ensure optimal retention of the mineral salts dissolved in the feed water, in order to obtain purified water of a very high quality.

Zubehörinformationen: Filtration panels
These filtration panels are accessories for demineralisation cylinders specifically designed for use in laboratories or health facilities. The cylinders ensure optimal retention of the mineral salts dissolved in the feed water, in order to obtain purified water of a very high quality.
With 2 or 3 filters.
One filtration panel comprises a flow control valve, a pressure regulator, an upstream pressure gauge, a 5 µm CA prefilter, a digital resistivity meter, a 1 µm final filter, a downstream pressure gauge and 2 bleed valves.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge