17284.14
17284.06
17284.14EA
195
EUR
InStock
17284.14
17284.06
Arsen(II)sulfid tech. 90%, technische Qualität
Arsen(II)sulfid
Purity: 90%
CAS No.: 12044-79-0
Molecular Formula: As2S2
Formula Weight: 213.97
Physical Form: Powder
Density: 3.5
Melting Point: ≈307°
Boiling Point: 565°
UN No.: 1557
MDL No.: MFCD00049841