Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

VWR® Vacuum Aspiration Systems

Lieferant: VWR Collection

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

181-0658EA 878 EUR InStock
181-0658 181-0659
VWR® Vacuum Aspiration Systems
Pumpen Vakuum-Absaugsysteme
The VWR® vacuum aspiration system is designed for laboratory waste recovery and the separation of liquids and solids.

  • Compact, space-saving, user-friendly design
  • Level detection system prevents bottle overfill (4 L)
  • Parts in contact with liquids are autoclavable
  • Contamination and protect personnel safety
  • Adjustable vacuum level with indicator
  • Collection bottle outlet with hydrophobic filter 0,45 μm prevents aerosol and liquid contamination
  • 15 ml/s aspiration rate

Designed with compact and space-saving housing for laboratory waste recovery and the separation of liquids and solids.

Package includes main unit, one bottle, one operating handle with tube, one single-needle, one long type needle, one 8-channel-needle, one tip detrusion adapter, and one 8-channel injector.

Zertifizierungen: EMC and LVD, UKCA certificated.

Bestellinformation: Two-year warranty.

Lieferumfang: VWR® 4 L and 2 L vacuum aspiration system basic unit is supplied with main unit, 4 L bottle for 4 L version and 2 L bottle for 2 L version, handle set with eight kinds of standard adapters, hydrophobic filter 0,45 μm. Components are also available separately.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge