Organic reference standard, Ponceau 4R, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
18137-25MG
18137-25MGEA 160 EUR InStock
18137-25MG
Organic reference standard, Ponceau 4R, analytical standard, Supelco®
Standards Organische Standards
Ponceau 4R is a synthetic red azo dye and is used as a food colorant. It is used in range of alcoholic and non-alcoholic beverages, food stuffs, preserved fruits and more. It exists as trisodium-2-hydroxy-1-(4-sulfonato-1-naphthylazo)-6,8-naphthalenedisulphonate.
Ponceau 4R was used in the determination of contents of several additives in different non-alcoholic beverages using HPLC. It was used in understanding the genotoxicity of colorants. It was also used in preparing a falsified drink in the analysis of methanol-ethanol mixture using biosensors amperometric system.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1d4663611bf1e52b2f757b0cc99b37930ddc49f&vid=f1d4663611bf1e52b2f757b0cc99b37930ddc49f&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge