Poly(Bisphenol A carbonate) analytical standards kit, molecular weight series, for GPC
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
181676-1SET
181676-1SETEA 468 EUR InStock
181676-1SET
Poly(Bisphenol A carbonate) analytical standards kit, molecular weight series, for GPC
Standards Gaschromatographie-Standards
Poly(Bisphenol A carbonate) (PC) is a thermoplastic material. It is injected in molding operations and is majorly studied to understand its thermal behaviour, especially thermal degradation property.
Kit components:
Average Mw 23000; average Mn 13,000 25 g
Average Mw 31000; average Mn 18,000 25 g
Average Mw 40000; average Mn 21,000 25 g
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d78d4d8d8658650f71c5d5c2ec3e83d28dab796a&vid=d78d4d8d8658650f71c5d5c2ec3e83d28dab796a&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge