Empty columns, HR series
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
19-7403-01
19-7403-01EA 902 EUR InStock
19-7403-01
Empty columns, HR series
Chromatographiesäulen
HR 16 columns are designed for high resolution liquid chromatography of biomolecules performed at low to medium pressures. The columns are suitable for use with high performance media such as Sepharose High Performance and SOURCE medium. Column hardware consists of borosilicate glass, ETFE, PP, PEEK, EPDM and PTFE.
  • Compatible with any high performance system including ÄKTA design systems
  • Provide precision sample application and elution and excellent physical and chemical stability
  • Glass construction, with no metal parts, allows visual observation at all times
  • Resistant to most solutions used in liquid chromatography except hydrocarbons, aromatic solvents and chlorinated hydrocarbons
  • Bed support: PP
Wide range of accessories, adapters and replacement parts available.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge