VWR® Protein Precipitation Kit
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1B1301-KITEA 185 EUR InStock
1B1301-KIT
VWR® Protein Precipitation Kit
Tests Proteintests
The Protein Precipitation Kit utilises a simple trichloroacetic acid (TCA) based method to precipitate proteins in complex mixtures. Only protein in the sample is pelleted, while interfering salts and surfactants remain in the supernatant. A gentle acetone wash removes TCA and co-precipitants allowing the end-user to recover the protein in a small volume of buffer.
  • Removal of interfering salts, detergents and buffers
  • Reproducible and consistent results
Bestellinformation: Kit contains: Protein precipitation kit Solution A; 25 ml, protein precipitation kit Solution B, 250 ml; and wash buffer, 125 ml.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge