VWR® Fluorescent DNA Quantitation Kit
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1B1558-KITEA 281 EUR InStock
1B1558-KIT
VWR® Fluorescent DNA Quantitation Kit
Tests Nukleinsäure-Tests
Fluorescent DNA Quantitation Kit uses fluorescent bisBenzimide H33258 dye to bind to AT sequences of the minor groove of double-stranded DNA. In the assay, the sample is excited at 360 nm and when the dye binds to dsDNA an emission spectrum at 460 nm is produced.
  • Highly specific quantitation of DNA
  • Sensitive to low nanogram per ml levels of DNA
  • Assay tolerates RNA and protein contaminants
Fluorescent DNA Quantitation Kit can be used to quantify dsDNA in the low nanogram per ml (10 ng/ml) range, up to 5 microgram per ml (5 µg/ml).
Lieferumfang: Kit includes 10X of Fluorescent DNA Assay Buffer (200 ml), Fluorescent DNA Dye (250 µl), and Fluorescent DNA Standard (1 ml).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge