Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MME Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SN5c]

Lieferant: Tonbo Biosciences

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

20-0108-T025 20-0108-T100 20-0108-T500
20-0108-T025EA 94.8 EUR InStock
20-0108-T025 20-0108-T100 20-0108-T500
Anti-MME Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SN5c]
Antikörper
The SN5c antibody is specific for human CD10, a 100 kDa type II transmembrane protein also known as Common Acute Lymphoblastic Leukemia Antigen (CALLA). In normal cells, it is expressed on early B and T lymphoid precursors, neutrophils and on various epithelia. CD10 is also expressed on several lymphoma cells including Burkitt's, acute lymphoblastic, and follicular germinal center lymphomas. CD10 functions as an endopeptidase and is involved in B cell development, as well as a mediator of neutrophil inflammatory responses.

Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: SN5c
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge