Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Bovine fibrinogen

Lieferant: H2B SAS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


2000-55-100ML 2000-55-500ML 2000-55-1L
2000-55-100MLEA 88 EUR InStock
2000-55-100ML 2000-55-500ML 2000-55-1L
Bovine fibrinogen
Proteine und Peptide
This ready-to-use 5% frozen solution enables a direct use of the product without further manipulation.

  • Total proteins: >35,0 g/L
  • Shelf life: 3 years
  • Sterile filtered 0,2 µm

SEQENS IVD provides highly purified fibrinogen manufactured according to Cohn fractionation method. The plasma used is collected from healthy animals of French origin. Bovine Fibrinogen is developed for RPF supplement, used with Baird-Parker agar, for the detection of Staphylococcus aureus.

Full traceability and batch-to-batch repeatability guaranteed.

Zertifizierungen: Bovine plasma used for the manufacture of this product is collected from French healthy cattle declared free from BSE disease in its clinical forms. It is sourced in EU-approved French slaughterhouses.

Lieferumfang: Supplied as a ready to use solution in aqueous saline buffer containing trisodium citrate and sodium chloride.


Order Now

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge