VWR®, Schliffhülsen für Normschliffe
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
201-0039EA 83.5 EUR InStock
201-0039 201-0040 201-0041 201-0042 201-0043 201-0044 201-0045 201-0046 201-0048
VWR®, Schliffhülsen für Normschliffe
Normschliff-Manschetten und Faltenbälge Schliffmanschetten
PTFE, mit Griffbund.
  • Very good chemical resistance
  • Gas, liquid and vacuum-tight
  • Temperature resistant from −200 to +280 °C
For fitting onto standard ground cones. Sturdy construction with protruding grip so sleeves can be detached easily. With sealing rings on the outside of the ground socket, which prevent ground joints from seizing and give a very good seal. Grease-free connection; no grease residue to contaminate sample.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge