Splash heads, vertical distillation, Quickfit®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SH7/12 SH7/13 SH7/53
201-0260EA 109 EUR InStock
201-0260 201-0261 201-0262
Splash heads, vertical distillation, Quickfit®
Destillationsbauteile Destillieraufsätze
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
A splash head for use in distillation assemblies in which the condenser is assembled in the vertical mode. Lower cone accepts flask. Upper cone accepts condenser. Splash head reduces entrainment of raw liquid into the vapour stream.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge