Druckhalteventil mit manueller Entlüftung, BOLA
Lieferant: Bohlender
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E683-18
201-0290EA 723 EUR InStock
201-0290
Druckhalteventil mit manueller Entlüftung, BOLA
Ventile Überdruckventile
Valve made from PPS with flow body made from PTFE. Can be used as a pressure relief valve with adjustable control of pressure or as a valve to prevent a fall in pressure during the filling process. Flow body with 2 GL 18 threaded connectors and valve with toggle for manual venting by tightening, set screw and lock nut to adjust and fix the application pressure in the range from 0,1 to 10 bar (factory setting 1,5 bar).
  • Very good chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −20 to +150°C
  • Pressure resistant up to a maximum of 10 bar, vacuum suitable
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge