Splash heads, anti-climb, Quickfit®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SH15/29 SH15/24 SH15/19
201-0573EA 143 EUR InStock
201-0573 201-0578 BIBBSH15/19
Splash heads, anti-climb, Quickfit®
Destillationsbauteile Destillieraufsätze
PYREX® borosilicate glass.
  • Protects against liquid ingress into the condenser
  • Useful for liquids that have tendency to foam
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge