Adapters with tube connector, straight or angled, Quickfit®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MF10/1B/SC MF10/2B/SC MF10/3B/SC MF10/4B/SC MF10/5B/SC MF13/2/SC MF13/3/SC MF13/4/SC
201-0567EA 28.3 EUR InStock
201-0567 201-0595 201-0568 201-0596 201-0645 201-3012 BIBBMF13/3/SC BIBBMF13/4/SC
Adapters with tube connector, straight or angled, Quickfit®
Adapter Adapter, Normschliff mit Schlaucholiven
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Screw thread connector accepts rubber hose with a bore of 9 mm
With straight or right angled connection for the attachment of rubber tubings etc. Detachable plastic screw thread connectors allow flexible tubing to be fitted easily and safely.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge