Microscale organic chemistry kit, Quickfit®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ML1 ML/FE10 ML/FE25 ML/FP10 ML/FP25 ML/FR10 ML/C1 ML/C2 ML/FR25 ML/MF23 ML/RA3 ML/SH4 ML/ST
BIBBML1EA 328 EUR InStock
BIBBML1 212-0618 212-0619 212-0620 212-0621 212-0622 201-3043 201-3036 212-0623 201-3061 271-0001 201-3007 201-3011
Microscale organic chemistry kit, Quickfit®
Mikrochemie-Bausätze
Microscale shemistry kit that is designed for organic chemistry experiments using milligram quantities of reagents. The components allow the construction of many assemblies including reflux, distillation, moisture sensitive reactions and fractional distillation.
  • Comprises 12 items of glassware that all feature 14/10 joint sizes for compatibility- see spare parts list
  • Other components compatible with the kit ar listed below under accessories
  • All components utilise a combination of standard Quickfit® ground glass joints together with screwthread couplings, sealing 'O' rings and plastic screw caps
  • The screwthread connector system eliminates the requirement for grease, saves on space, using clamps and vastly reduces the danger of glassware becoming loose during reactions
Lieferumfang: Supplied in a sturdy Gratnells tray with protective foam insert. Also included in the kit is a bag of spare caps, 'O' rings, sealing discs.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge