Joint clips for conical joints, POM, Quickfit®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
KC34 KC10 KC12 KC14 KC19 KC24 KC29 KC45 KC40
201-8570EA 77.3 EUR InStock
201-8570 201-8185 201-8186 201-8566 201-8567 201-8568 201-8569 201-8192 201-3032
Joint clips for conical joints, POM, Quickfit®
Klemmen Schliffklemmen
POM, coloured. Polyacetal clips for retaining glassware having Quickfit® conical joints.
  • Easy to fit and remove
  • Can be used at temperatures up to +80 °C
  • Clips are colour coded for easy identification
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge